HF Scientific™ ProCal™ Turbidity Standard Reagents Kit for Hach™ 2100Q Portable Turbidimeter DFS Item
Standards not harmful to the environment or to worker safety
Supplier:  HF Scientific™ HF52570
 View more versions of this product
Catalog No. 15-393-643
Explore available promotions
Description
Description
  • Pre-diluted
  • Do not include formazin
  • Preservative used enhances stability
  • Biodegradable, safe and non-toxic
  • Ready and easy to use
  • Factory certified by HF scientific
  • Custom NTU produced
  • Available in different size bottles
Specifications
Specifications
Primary Turbidity Standard
800 NTU
8.45 oz.
Hach 2100Q Portable Turbidimeter
EPA
250 mL