missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CoorsTek™ High-Form Alumina Crucible Covers

Flat flanged style

Supplier:  CoorsTek™ 65611

 View more versions of this product

Catalog No. 12-220-2C


Explore available promotions

Description

Description

Flange holds covers securely in place

Order Info

Thickness: 0.125 in. (3.2mm)

Specifications

Specifications

Alumina
Cover for 50 mL high-form crucible 65611
Videos
SDS
Documents

Documents

Promotions

Promotions