missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
CoorsTek™ High-Form Alumina Crucible Covers
Flat flanged style
$162.13 - $302.77
Catalog Number | Mfr. No. | For Use With (Application) | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Application) | Price | Quantity | |||||
12-220-2D
|
CoorsTek™
65613 |
Cover for 100 mL high-form crucible 65613 |
Each for $302.77
|
|
|||||
08-226-16A
|
CoorsTek™
CRS65602 |
Cover for 10 mL high-form crucible 65533 |
Each for $162.13
|
|
|||||
12-220-2C
|
CoorsTek™
65611 |
Cover for 50 mL high-form crucible 65611 |
Each for $274.93
|
|
|||||
12-220-2A
|
CoorsTek™
65605 |
Cover for 10mL high-form crucible 65605 |
Each for $220.57
|
|
|||||
12-220-2B
|
CoorsTek™
65608 |
Cover for 20 mL high-form crucible 65608 |
Each for $239.77
|
|
|||||
Description
Flange holds covers securely in place
Order Info
Thickness: 0.125 in. (3.2mm)
Specifications
Alumina |