missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
CoorsTek™ High-Form Alumina Crucible Covers
Flat flanged style
Supplier: CoorsTek™ 65613
Description
Flange holds covers securely in place
Order Info
Thickness: 0.125 in. (3.2mm)
Specifications
Alumina | |
Cover for 100 mL high-form crucible 65613 |