Learn More
High-Frequency Tesla Coil
Safe source of high-frequency, high-voltage electricity.
$43.69 - $1255.26
Specifications
For Use With (Application) Transmit electricity without wires; Demonstrates fundamental principles of high frequency electrical phenomena
View More Specs
Products 2
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
S68853
View Documents
Customer Channels Group (Ccg)
10212
Plasma Tube Only
Each for $43.69
Only null left
S43505X
Accessories View Documents
Customer Channels Group (Ccg)
10206
Tesla Coil
Each for $1,255.26
Only null left
Description
Description
Transmit electricity without wires. Plasma tube for Tesla coil available separately.
  • Arcs to 3 in. (7.6cm)
  • 50,000V potential
  • Fully assembled coil
Specifications
Specifications
Transmit electricity without wires; Demonstrates fundamental principles of high frequency electrical phenomena
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'