missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cytiva HiScreen™ MabSelect™, Xtra™, SuRe™, SuRe LX™ Prepacked Columns
Designed for method optimization and parameter screening for capture of monoclonal antibodies. Cytiva HiScreen™ MabSelect™, Xtra™, SuRe™, SuRe LX™ Prepacked Columns come ready-to-use for convenience.
$1635.86 - $2145.44
Specifications
Buffer | 8M Urea, 6M Guanidine Hydrochloride, 2% Benzyl Alcohol or 20% Ethanol |
---|---|
Includes | Stoppers at the inlet and outlet |
Kit Size | 1 x 4.7mL |
Bed Support | Filter |
Diameter (English) Inner | 0.3 in. |
Catalog Number | Mfr. No. | Ligand | Product Type | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Ligand | Product Type | Price | Quantity | ||||
45-002-526
|
Cytiva
28926973 |
Recombinant Protein A (E.coli) | HiScreen MabSelect Prepacked Column |
Each for $1,635.86
|
|
||||
45-002-528
|
Cytiva
28926977 |
Alkali-stabilized Protein A-derived (E. coli) | HiScreen MabSelect SuRe Prepacked Column |
Each for $2,145.44
|
|
||||
45-002-525
|
Cytiva
17547415 |
Alkali-stabilized Protein A-derived (E. coli) | HiScreen MabSelect SuRe LX Prepacked Column |
Each for $2,135.90
|
|
||||
Description
- Ready-to-use columns for convenient process development
- Rigid, highly cross-linked agarose matrix
- Bed height of 10cm is designed to allow method optimization and parameter screening
- Easily connected in series to achieve 20cm bed height
- Small bed volume (4.7mL) gives fast result and minimal sample/buffer consumption
- Reproducible results, scalable to BioProcess columns packed with the same chromatography media using the same linear fluid velocity
MabSelect
- For capture of MAbs from large sample volumes
- Prepacked with protein A-derived media
- Compatible with high flow rates and high pressure when scaling up
- Increased binding capacity enables purification of MAbs from high-level expression feedstock
- Also optimized for Fc-fusion proteins
- Very low, unspecific binding due to high ligand selectivity and matrix hydrophilicity
- Prepacked with protein A-derived media
- With novel, alkali-tolerant recombinant protein A ligand that provides greater stability and allows use of 0.1 to 0.5M sodium hydroxide for cleaning-in-place (CIP)
- Enhanced protease resistance resulting in lower ligand leakage
- High dynamic binding capacity
Specifications
8M Urea, 6M Guanidine Hydrochloride, 2% Benzyl Alcohol or 20% Ethanol | |
1 x 4.7mL | |
0.3 in. | |
3.93 in. | |
Polypropylene | |
Column |
Stoppers at the inlet and outlet | |
Filter | |
0.77cm | |
Epoxy Activation | |
2° to 8°C |