Cytiva HiTrap MabSelect PrismA Column
Protein A Column
Supplier:  Cytiva 17549854
 View more versions of this product
Catalog No. 09928071
Explore available promotions
Description
Description
These columns are prepacked with MabSelect PrismA protein A chromatography resin that has been improved with an optimized high-flow agarose base matrix and a genetically engineered protein A-derived ligand, allowing efficient cleaning between purification runs.
Specifications
Specifications
5 mL
5 bar (0.5 MPa, 70 psi)
Antibody affinity chromatography
<20 mL/min.
5 x 5 mL
Pre-packed Column