missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cytiva HiTrap MabSelect PrismA Column
Protein A Column
$310.48 - $5520.92
Specifications
Max. Operating Pressure | 5 bar (0.5 MPa, 70 psi) |
---|---|
For Use With (Application) | Antibody affinity chromatography |
Type | Pre-packed Column |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
09928068
|
Cytiva
17549851 |
1 x 1 mL |
Each for $310.48
|
|
|||||
09928069
|
Cytiva
17549852 |
5 x 1 mL |
Each for $1,210.54
|
|
|||||
09928070
|
Cytiva
17549853 |
1 x 5 mL |
Each for $1,239.63
|
|
|||||
09928071
|
Cytiva
17549854 |
5 x 5 mL |
Each for $5,520.92
|
|
|||||
Description
These columns are prepacked with MabSelect PrismA protein A chromatography resin that has been improved with an optimized high-flow agarose base matrix and a genetically engineered protein A-derived ligand, allowing efficient cleaning between purification runs.
Specifications
5 bar (0.5 MPa, 70 psi) | |
Antibody affinity chromatography | |
Pre-packed Column |