Epredia™ HM 325 Rotary Microtome
Cut high-quality paraffin sections without a power supply with the robust Epredia™ HM 325 Rotary Microtome.
Supplier:  Epredia™ SH902100
 View more versions of this product
Catalog No. 23-900-667
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
  • One-handed zero orientation head
  • Quick-trim button
  • Section counter
  • Guide-rail guidance for the knife holder
  • Large, easy-to-clean waste tray extends below the blade holder
Ordering Information:
  • Blade holder/Knife holder must be ordered separately unless purchased as a package.
Includes:
  • Standard tools
  • Brushed aluminum cover plate (additional customized cover plates available)
  • Dust cover
  • Instruction manual
  • 902100A also includes a universal cassette clamp (715020S) and a disposable blade carrier E (705800)
  • 902100ER also includes a universal cassette clamp (715020S) and a disposable blade carrier ER (705830)
Specifications
Specifications
HM 325 Rotary Microtome
N/A
49 cm
28 cm
42 cm
23 kg
Rotary Microtome
19 in.
11 in.
16.5 in.
51 lb.