Honeywell™ North™ Silver Shield™ Chemical Resistant Aprons
Thin, lightweight aprons are available with and without sleeves.
Supplier: Honeywell™ SSCAS
Description
- Five layer PE and EVOH constructions provides excellent chemical resistant to over 280 different chemicals including alchohols, aliphatics, aromatice, chlorines, ketones and esters
- Provides high performance protection in the most demanding applications
Specifications
Coat Apron | |
Composite Fabric | |
Sleeves |
Small | |
Silver |