Honeywell™ North™ Silver Shield™ Chemical Resistant Aprons
Thin, lightweight aprons are available with and without sleeves.
$58.69 - $1472.02
Specifications
Material Composite Fabric
Color Silver
View More Specs
Products 4
Catalog Number Mfr. No. Size Type Price Quantity  
Catalog Number Mfr. No. Size Type Price Quantity  
29-635-972
View Documents
Honeywell™
SSA
Universal Chemical-resistant Apron
Each for $58.69
19-088-1683D
View Documents
Honeywell™
SSCAXL
X-Large Coat Apron
Each for $121.41
Case of 10 Each for $370.10
19-150-0896
View Documents
Honeywell™
SSCAS
Small Coat Apron
Case of 10 Each for $1,363.32
19-150-0897
View Documents
Honeywell™
SSCAM
Medium Coat Apron
Case of 10 Each for $1,472.02
Description
Description
  • Five layer PE and EVOH construction provides excellent chemical resistant to over 280 different chemicals including alchohols, aliphatics, aromatice, chlorines, ketones and esters
  • Provides high performance protection in the most demanding applications
Specifications
Specifications
Composite Fabric
Silver
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'