missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cytiva HyClone™ Cosmic Calf™ Serum (U.S.)
Contains high quality iron-supplemented bovine calf serum with added growth promoting factors.
$48.56 - $390.88
Specifications
Certifications/Compliance | Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities |
---|---|
Content And Storage | Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C. |
Sterility | Sterile |
Sterilization Method | Sequential-filtered, 100 nm, Gamma-irradiated |
Catalog Number | Mfr. No. | Quantity | Processing | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Processing | Price | Quantity | |||||
SH3008702
|
Cytiva
SH3008702 |
100 mL | None |
Each for $48.56
|
|
|||||
SH3008703
|
Cytiva
SH3008703 |
500 mL | None |
Each for $213.65
|
|
|||||
SH3008704
|
Cytiva
SH3008704 |
1000 mL | None |
Each for $390.88
|
|
|||||
SH3008703HI
|
Cytiva
SH3008703HI |
500 mL | Heat Inactivated |
Each for $267.06
|
|
|||||
Description
- Age at time of collection: typically 16 - 22 weeks
- Iron-supplemented with additional growth promoting components
Specifications
Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities | |
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C. | |
Sterile | |
Sequential-filtered, 100 nm, Gamma-irradiated |