missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cytiva HyClone™ Cosmic Calf™ Serum (U.S.)

Contains high quality iron-supplemented bovine calf serum with added growth promoting factors.

Supplier:  Cytiva SH3008703

 View more versions of this product

Catalog No. SH3008703


Explore available promotions

Description

Description

  • Age at time of collection: typically 16 - 22 weeks
  • Iron-supplemented with additional growth promoting components
Specifications

Specifications

Albumin, Iron, Lactate Dehydrogenase, Calcium, Chloride, pH, Gamma Globulin, Potassium, SGOT, Insulin, Sodium, Alkaline Phosphatase, TIBC, Total Bilirubin, Total Protein, Blood Urea Nitrogen, Calcium, Chloride, Creatinine, Gamma Globulin, Glucose, IgG, Insulin
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C.
Iron and naturally derived components
500 mL
Sequential-filtered, 100 nm, Gamma-irradiated
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
Calf Serum; Cosmic Calf; 500 mL
Calf Bovine Serum
Sterile
None
Videos
SDS
Documents

Documents

Promotions

Promotions