missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cytiva HyClone™ Donor Equine Serum (U.S.)
Supplement cell culture growth with U.S. sourced, collected from selected donor herds that receive regular veterinary inspection and care.
$34.50 - $147.06
Specifications
Additional Assays | Albumin, Lactate Dehydrogenase, Osmolality, Percent Saturation, Chloride, Creatinine, SGOT, SGPT, Inorganic Phosphorus, Iron, Total Bilirubin, Alkaline Phosphatase, Total Protein, Blood Urea Nitrogen, Calcium, Chloride, Creatinine, Gamma Globulin, Glucose, Inorganic Phosphorus, Iron |
---|---|
Country of Origin | United States |
Form | Liquid |
Product Type | Equine Serum |
Sterilization Method | Sequential Triple-filtered, 100 nm |
Catalog Number | Mfr. No. | Description | Quantity | Processing | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Quantity | Processing | Price | Quantity | |||
SH3007402
|
Cytiva
SH3007402 |
Donor Equine Serum (U.S.) | 100 mL | None |
Each for $34.50
|
|
|||
SH3007403
|
Cytiva
SH3007403 |
Donor Equine Serum (U.S.) | 500 mL | None |
Each for $147.06
|
|
|||
Description
- Format: Liquid
Specifications
Albumin, Lactate Dehydrogenase, Osmolality, Percent Saturation, Chloride, Creatinine, SGOT, SGPT, Inorganic Phosphorus, Iron, Total Bilirubin, Alkaline Phosphatase, Total Protein, Blood Urea Nitrogen, Calcium, Chloride, Creatinine, Gamma Globulin, Glucose, Inorganic Phosphorus, Iron | |
Liquid | |
Sequential Triple-filtered, 100 nm |
United States | |
Equine Serum | |
BVDV, Cytopathogenic Agents, Rabies Virus, Reovirus |