missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cytiva HyClone™ Donor Equine Serum (U.S.)

Supplement cell culture growth with U.S. sourced, collected from selected donor herds that receive regular veterinary inspection and care.

Supplier:  Cytiva SH3007402

 View more versions of this product

Catalog No. SH3007402


Explore available promotions

Description

Description

  • Format: Liquid
Specifications

Specifications

Albumin, Lactate Dehydrogenase, Osmolality, Percent Saturation, Chloride, Creatinine, SGOT, SGPT, Inorganic Phosphorus, Iron, Total Bilirubin, Alkaline Phosphatase, Total Protein, Blood Urea Nitrogen, Calcium, Chloride, Creatinine, Gamma Globulin, Glucose, Inorganic Phosphorus, Iron
United States
Equine Serum
Sequential Triple-filtered, 100 nm
BVDV, Cytopathogenic Agents, Rabies Virus, Reovirus
Donor Equine Serum (U.S.)
Liquid
100 mL
None
No
Videos
SDS
Documents

Documents

Promotions

Promotions