missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cytiva HyClone™ Dulbecco's Phosphate Buffered Saline, Solution
Buffer cell culture with PBS in solution.
Supplier: Cytiva SH3002804
Description
- Format: Liquid
Specifications
Manufactured under cGMP and ISO 9001 certified facilities | |
HyClone Dulbecco's Phosphate Buffered Salines (DPBS) Without Calcium, Magnesium, Phenol Red | |
Liquid | |
Sterile | |
None | |
≤0.025EU/mL | |
276±10mOsm/kg H2O | |
20 L |
15° to 30°C | |
1 X | |
Dulbecco's Phosphate Buffered Saline | |
0.1 μm Sterile-filtered | |
No Calcium, No Magnesium | |
Serves as an irrigating, transporting or diluting fluid while maintaining cell tonicity and viability for a limited period of time | |
7 to 7.6 | |
RUO |