missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cytiva HyClone™ Dulbecco's Phosphate Buffered Saline, Solution
Buffer cell culture with PBS in solution.
$8.90 - $692.46
Specifications
Certifications/Compliance | Manufactured under cGMP and ISO 9001 certified facilities |
---|---|
Content And Storage | 15° to 30°C |
Form | Liquid |
Product Type | Dulbecco's Phosphate Buffered Saline |
Sterility | Sterile |
Catalog Number | Mfr. No. | Quantity | Concentration | With Additives | Without Additives | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Concentration | With Additives | Without Additives | Price | Quantity | |||
SH3002801
|
Cytiva
SH3002801 |
100 mL | 1 X | None | No Calcium, No Magnesium, No Phenol Red |
Each for $8.90
|
|
|||
SH30028LS
|
Cytiva
SH30028LS |
6 x 1000 mL | 1 X | None | No Calcium, No Magnesium, No Phenol Red |
Pack of 6 Each for $123.63
|
|
|||
SH3026402
|
Cytiva
SH3026402 |
1000 mL | 1 X | Calcium, Magnesium | No Phenol Red |
Each for $28.73
|
|
|||
SH3002808
|
Cytiva
SH3002808 |
10 L | 1 X | None | No Calcium, No Magnesium |
Each for $532.42
|
|
|||
SH30028FS
|
Cytiva
SH30028FS |
6 x 500 mL | 1 X | None | No Calcium, No Magnesium, No Phenol Red |
Pack of 6 Each for $81.76
|
|
|||
SH30264FS
|
Cytiva
SH30264FS |
6 x 500 mL | 1 X | Calcium, Magnesium | No Phenol Red |
Pack of 6 Each for $75.31
|
|
|||
SH3002803
![]() |
Cytiva
SH3002803 |
1000 mL | 1 X | None | No Calcium, No Magnesium, No Phenol Red |
Each for $22.88
|
![]() |
|||
SH3026401
|
Cytiva
SH3026401 |
500 mL | 1 X | Calcium, Magnesium | No Phenol Red |
Each for $17.24
|
|
|||
SH3002802
![]() |
Cytiva
SH3002802 |
500 mL | 1 X | None | No Calcium, No Magnesium, No Phenol Red |
Each for $14.16
|
![]() |
|||
SH3037803
|
Cytiva
SH3037803 |
1 L | 10 X | None | No Calcium, No Magnesium, No Phenol Red |
Each for $62.96
|
|
|||
SH3037802
|
Cytiva
SH3037802 |
500 mL | 10 X | None | No Calcium, No Magnesium, No Phenol Red |
Each for $35.71
|
|
|||
SH3002804
|
Cytiva
SH3002804 |
20 L | 1 X | None | No Calcium, No Magnesium |
Each for $692.46
|
|
|||
Description
- Format: Liquid
Specifications
Manufactured under cGMP and ISO 9001 certified facilities | |
Liquid | |
Sterile | |
≤0.025EU/mL | |
276±10mOsm/kg H2O | |
RUO |
15° to 30°C | |
Dulbecco's Phosphate Buffered Saline | |
0.1 μm Sterile-filtered | |
Serves as an irrigating, transporting or diluting fluid while maintaining cell tonicity and viability for a limited period of time | |
7 to 7.6 |