missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cytiva HyClone™ Dulbecco's Phosphate Buffered Saline, Solution

Buffer cell culture with PBS in solution.

Supplier:  Cytiva SH30028FS

 View more versions of this product

Catalog No. SH30028FS


Explore available promotions

Description

Description

  • Format: Liquid
Specifications

Specifications

Manufactured under cGMP and ISO 9001 certified facilities
HyClone Dulbecco's Phosphate Buffered Salines (DPBS) Without Calcium, Magnesium, Phenol Red
Liquid
Sterile
None
≤0.025EU/mL
276±10mOsm/kg H2O
6 x 500 mL
15° to 30°C
1 X
Dulbecco's Phosphate Buffered Saline
0.1 μm Sterile-filtered
No Calcium, No Magnesium, No Phenol Red
Serves as an irrigating, transporting or diluting fluid while maintaining cell tonicity and viability for a limited period of time
7 to 7.6
RUO
Videos
SDS
Documents

Documents

Promotions

Promotions