missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cytiva HyClone™ Fetal Bovine Serum (U.S.), Charcoal/Dextran Treated
Augment growth of cells with U.S.-sourced HyClone™ Charcoal/Dextran Treated Fetal Bovine Serum (FBS), containing reduced levels of many hormones and growth factors.
$291.40 - $1265.92
Specifications
Country of Origin | United States |
---|---|
Specialty Type | Charcoal/Dextran filtered |
Sterility | Sterile |
Grade | Specialty filtered |
Endotoxin Concentration | ≤ 10 EU/mL |
Catalog Number | Mfr. No. | Quantity | Processing | Heat Inactivated | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Processing | Heat Inactivated | Price | Quantity | |||
SH3006803
|
Cytiva
SH3006803 |
500 mL | None | No |
Each for $1,221.63
|
|
|||
SH3006803HI
|
Cytiva
SH30068.03HI |
500 mL | Heat Inactivated | Yes |
Each for $1,265.92
|
|
|||
SH3006802HI
|
Cytiva
SH3006802HI |
100 mL | Heat Inactivated | Yes |
Each for $329.29
|
|
|||
SH3006802
|
Cytiva
SH3006802 |
100 mL | None | No |
Each for $291.40
|
|
|||
Description
- Processed using a proprietary charcoal/dextran treatment that has been shown to reduce the levels of many hormones and growth factors
- This serum is assayed for a variety of components before and after treatment to ensure the efficacy of the treatment
- Charcoal/dextran treatment has been shown to be effective in reducing steroid levels for use of the serum in receptor studies or estrogen-related investigations
- Other studies indicate that charcoal/dextran treatments may be used to minimize lot-to-lot serum variability
- Filtered through three sequential 100 nm (0.1 μm) pore size-rated filters
- Before dispensing, each lot of serum is pooled using true pool technology to ensure uniformity and consistency between bottles
Specifications
United States | |
Sterile | |
≤ 10 EU/mL | |
No |
Charcoal/Dextran filtered | |
Specialty filtered | |
≤ 20 mg/dL | |
USDA Inspected |