missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cytiva HyClone™ Fetal Bovine Serum (U.S.), Charcoal/Dextran Treated

Augment growth of cells with U.S.-sourced HyClone™ Charcoal/Dextran Treated Fetal Bovine Serum (FBS), containing reduced levels of many hormones and growth factors.

Supplier:  Cytiva SH3006803

 View more versions of this product

Catalog No. SH3006803


Explore available promotions

Description

Description

  • Processed using a proprietary charcoal/dextran treatment that has been shown to reduce the levels of many hormones and growth factors
  • This serum is assayed for a variety of components before and after treatment to ensure the efficacy of the treatment
  • Charcoal/dextran treatment has been shown to be effective in reducing steroid levels for use of the serum in receptor studies or estrogen-related investigations
  • Other studies indicate that charcoal/dextran treatments may be used to minimize lot-to-lot serum variability
  • Filtered through three sequential 100 nm (0.1 μm) pore size-rated filters
  • Before dispensing, each lot of serum is pooled using true pool technology to ensure uniformity and consistency between bottles
Specifications

Specifications

1, 25 Dihydroxy Vitamin D, 17 Beta Estradiol, Calcium, Alkaline Phosphatase, Chloride, B12, Cortisol, Creatinine, Biotin, Folate, 25 Dihydroxy Vitamin D, FSH, Glucose, IgG, Inorganic Phosphorus, Insulin, Lactate Dehydrogenase, LH, Niacin, Potassium, Progesterone, 17 Beta Estradiol, Retinol, SGOT, SGPT, Sodium, T3, T4, Testosterone, Tocopherol, Total Bilirubin, Total Protein, Albumin, TSH, Vitamin C, Alkaline Phosphatase, B1, B12, B2, B6, Biotin
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C.
United States
500 mL
Sequential-filtered, 100 nm
None
≤ 10 EU/mL
≤ 20 mg/dL
USDA Inspected
EMEA Certified
Charcoal/Dextran Treated Fetal Bovine Serum, US Origin, None
Charcoal/Dextran filtered
Sterile
Specialty filtered
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
No
No
SDS
Documents

Documents

Promotions

Promotions