missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cytiva HyClone™ Phosphate Buffered Saline (PBS)
Buffer cell culture growth with PBS
$14.57 - $133.92
Specifications
Certifications/Compliance | cGMP and ISO 9001:2000 Guidelines |
---|---|
Content And Storage | 15° to 30°C |
Form | Liquid |
Product Type | Phosphate Buffered Saline (PBS) |
Sterilization Method | 0.1 μm Sterile-filtered |
Catalog Number | Mfr. No. | Quantity | Concentration | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Concentration | Price | Quantity | ||||
SH3025602
|
Cytiva
SH3025602 |
1000 mL | 1 X |
Each for $24.16
|
|
||||
SH3025601
|
Cytiva
SH3025601 |
500 mL | 1 X |
Each for $14.57
|
|
||||
SH30256LS
|
Cytiva
SH30256LS |
6 x 1000 mL | 1 X |
Pack of 6 Each for $133.92
|
|
||||
SH3025801
|
Cytiva
SH3025801 |
500 mL | 10 X |
Each for $33.29
|
|
||||
SH3025802
|
Cytiva
SH3025802 |
1000 mL | 10 X |
Each for $49.76
|
|
||||
SH30256FS
|
Cytiva
SH30256FS |
6 x 500 mL | 1 X |
Pack of 6 Each for $83.55
|
|
||||
Description
- QC-tested solution
Specifications
cGMP and ISO 9001:2000 Guidelines | |
Liquid | |
0.1 μm Sterile-filtered | |
=1.0EU/mL |
15° to 30°C | |
Phosphate Buffered Saline (PBS) | |
No Calcium, No Magnesium, No Phenol Red | |
7.0 to 7.2 |