Cytiva HyClone™ Phosphate Buffered Saline (PBS)
Buffer cell culture growth with PBS
Supplier:  Cytiva SH3025601
 View more versions of this product
Catalog No. SH3025601
Explore available promotions
Description
Description
  • QC-tested solution
Specifications
Specifications
cGMP and ISO 9001:2000 Guidelines
1 X
Phosphate Buffered Saline (PBS)
No Calcium, No Magnesium, No Phenol Red
7.0 to 7.2
15° to 30°C
Liquid
0.1 μm Sterile-filtered
=1.0EU/mL
500 mL