missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cytiva HyClone™ Trypsin Protease

Derived from porcine pancreas. HyClone™ Trypsin Protease is used to disaggregate adherent cells from substrate or surface to which they are attached.

$16.44 - $61.68

Specifications

Content And Storage Store frozen
Form Liquid
Sterilization Method 0.1 μm Sterile-filtered
Source Porcine pancreas
View More Specs

Products 6
Catalog Number Mfr. No. Quantity Formulation Without Additives Price Quantity  
Catalog Number Mfr. No. Quantity Formulation Without Additives Price Quantity  
SV3003101
View Documents
Cytiva
SV3003101
100 mL 2.5 g porcine trypsin (1:250/L gamma irradiated) in HBSS with 1 g/L EDTA No Calcium, No Magnesium
Each for $38.34
 
SV3003701
View Documents
Cytiva
SV3003701
100 mL 25.0 g porcine trypsin (1:250/L) in HBSS No Calcium, No Magnesium, No EDTA, No Phenol Red
Each for $61.68
 
SH3023601
View Documents Promotion Details
Cytiva
SH3023601
100 mL 0.5 g porcine trypsin (1:250/L gamma irradiated) in HBSS with 0.2 g/L EDTA No Calcium, No Magnesium
Each for $16.44
 
SH3004201
View Documents
Cytiva
SH3004201
100 mL 2.5 g porcine trypsin (1:250/L gamma irradiated) in HBSS with 0.2 g/L EDTA No Calcium, No Magnesium
Each for $17.68
 
SH3004202
View Documents
Cytiva
SH3004202
500 mL 2.5 g porcine trypsin (1:250/L gamma irradiated) in HBSS with 0.2 g/L EDTA No Calcium, No Magnesium
Each for $60.50
 
SH3023602
View Documents
Cytiva
SH3023602
500 mL 0.5 g porcine trypsin (1:250/L gamma irradiated) in HBSS with 0.2 g/L EDTA No Calcium, No Magnesium
Each for $58.08
 
Description

Description

    • Derived from porcine pancreas
    • Gamma irradiated prior to hydration and filling
    • Formulated without calcium and magnesium
Specifications

Specifications

Store frozen
Liquid
0.1 μm Sterile-filtered
Porcine pancreas
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'