missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cytiva HyClone™ Trypsin Protease
Derived from porcine pancreas. HyClone™ Trypsin Protease is used to disaggregate adherent cells from substrate or surface to which they are attached.
Supplier: Cytiva SH3023602
Description
- Derived from porcine pancreas
- Gamma irradiated prior to hydration and filling
- Formulated without calcium and magnesium
Specifications
Store frozen | |
Liquid | |
Trypsin, porcine, Hanksâ Balanced Salt solution (HBSS), EDTA | |
0.5 g porcine trypsin (1:250/L gamma irradiated) in HBSS with 0.2 g/L EDTA | |
Porcine pancreas |
0.05% (1 X) | |
0.1 μm Sterile-filtered | |
No Calcium, No Magnesium | |
500 mL |