missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
HYDRANAL™ - Water Standard 1.0, Standard for Karl Fischer titration (water content 1 mg/g = 0.1%), verified against NIST SRM 2890 & NMIJ CRM 4222, Honeywell™ Fluka™
Supplier: Honeywell Chemicals 348286X40ML
Specifications
1.000 g/cm3 | |
1 Kit | |
Standard for Karl Fischer titration (water content 1 mg/g = 0.1%), exact value on report of analysis | |
≥90.0000%,≥1.0000% | |
153°C to 155°C | |
C7H8O | |
MFCD00007941 | |
Methoxybenzene, Methyl phenyl ether | |
COC1=CC=CC=C1 | |
108.14 | |
CHEBI:16579 |
43°C | |
HYDRANAL™ - Water Standard 1.0 | |
100-66-3,108-32-7 | |
<100.0000%,<10.0000% | |
Ampoule | |
HCONH2 | |
UN2222 | |
RDOXTESZEPMUJZ-UHFFFAOYSA-N | |
anisole | |
7519 | |
Karl Fischer |
Chemical Identifiers
100-66-3 | |
108.14 | |
RDOXTESZEPMUJZ-UHFFFAOYSA-N | |
7519 | |
anisole |
C7H8O | |
MFCD00007941 | |
Methoxybenzene, Methyl phenyl ether | |
CHEBI:16579 | |
COC1=CC=CC=C1 |
Safety and Handling
ShelfLife : 1800 days from date of manufacture
DOTInformation : Transport Hazard Class: 3; Packing Group:3: III; Proper Shipping Name: Anisole solution
Recommended Storage : Room Temp