missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
HYDRANAL™ - Water Standard 1.0, Standard for Karl Fischer titration (water content 1 mg/g = 0.1%), verified against NIST SRM 2890 & NMIJ CRM 4222, Honeywell™ Fluka™
$251.38 - $251.38
Chemical Identifiers
CAS | 100-66-3 |
---|---|
Molecular Formula | C7H8O |
Molecular Weight (g/mol) | 108.14 |
MDL Number | MFCD00007941 |
InChI Key | RDOXTESZEPMUJZ-UHFFFAOYSA-N |
Synonym | Methoxybenzene, Methyl phenyl ether |
PubChem CID | 7519 |
ChEBI | CHEBI:16579 |
IUPAC Name | anisole |
SMILES | COC1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
60050007
|
Honeywell Chemicals
348286X40ML |
1 Kit |
Each for $251.38
Case of 6 Each for $1,508.28
|
|
|||||
Chemical Identifiers
100-66-3 | |
108.14 | |
RDOXTESZEPMUJZ-UHFFFAOYSA-N | |
7519 | |
anisole |
C7H8O | |
MFCD00007941 | |
Methoxybenzene, Methyl phenyl ether | |
CHEBI:16579 | |
COC1=CC=CC=C1 |
Specifications
1.000 g/cm3 | |
1 Kit | |
≥90.0000%,≥1.0000% | |
153°C to 155°C | |
C7H8O | |
MFCD00007941 | |
Methoxybenzene, Methyl phenyl ether | |
COC1=CC=CC=C1 | |
108.14 | |
CHEBI:16579 | |
HYDRANAL™ - Water Standard 1.0, Standard for Karl Fischer titration (water content 1 mg/g = 0.1%), exact value on report of analysis |
43°C | |
100-66-3,108-32-7 | |
<100.0000%,<10.0000% | |
Ampoule | |
HCONH2 | |
UN2222 | |
RDOXTESZEPMUJZ-UHFFFAOYSA-N | |
anisole | |
7519 | |
Karl Fischer |
Safety and Handling
ShelfLife : 1800 days from date of manufacture
DOTInformation : Transport Hazard Class: 3; Packing Group:3: III; Proper Shipping Name: Anisole solution
Recommended Storage : Room Temp