missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Hydrion™ pH Test Papers, Wide-Range
Select the pH range you need, quickly and inexpensively.
$31.87 - $52.10
Specifications
For Use With (Application) | To determine pH within half unit via distinct color changes |
---|---|
Includes | Color comparison scale |
Catalog Number | Mfr. No. | pH Range | Type | Quantity | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | pH Range | Type | Quantity | Price | Quantity | |||
S60150
|
Customer Channels Group (Ccg)
MES165A |
2 to 10 | Type A Vial | 10 Pack |
Pack of 10 Each for $52.10
|
|
|||
S60152
|
Customer Channels Group (Ccg)
MES165B |
1 to 11 | Type B pH Paper | 10 Pack |
Pack of 10 Each for $52.10
|
|
|||
S60144
|
Customer Channels Group (Ccg)
150EMD |
1 to 11 | Hydrion™ D/R AB Dispenser | Each |
Each for $31.87
|
|
|||
Description
- Type A indicates pH 2, 4, 6, 8 and 10
- Type B indicates pH 1, 3, 5, 7, 9 and 11
Order Info
- Cat. No. S60146 and S60148: 5 rolls and 2 color charts per kit, 10 kits per case
- Cat. No. S60152: 10 vials per box
Specifications
To determine pH within half unit via distinct color changes | |
Color comparison scale |