Hydrion™ pH Test Papers, Wide-Range
Select the pH range you need, quickly and inexpensively.
Supplier:  Customer Channels Group (Ccg) MES165A
 View more versions of this product
Catalog No. S60150
Explore available promotions
Description
Description
  • Type A indicates pH 2, 4, 6, 8 and 10
  • Type B indicates pH 1, 3, 5, 7, 9 and 11
Order Info
  • Cat. No. S60146 and S60148: 5 rolls and 2 color charts per kit, 10 kits per case
  • Cat. No. S60152: 10 vials per box
Specifications
Specifications
2 to 10
Color comparison scale
Type A Vial
To determine pH within half unit via distinct color changes
10 Pack
Product Suggestions
Product Suggestions