missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hydrochloric acid, ACS reagent, ca. 37% solution in water, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 423795001

 View more versions of this product

Catalog No. AC423795001


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Hydrochloric acid
-35.0°C
57.0°C
1ppm max.
0.0005% max.
500 mL
ClH
HCl
1789
15, 4818
Solubility in water: miscible: 823g/L (0°C); 561g/L (60°C). Other solubilities: soluble in alcohol, benzene and ether, insoluble in hydrocarbons
Cl
36.46
CHEBI:17883
36.5 to 38.0%
ca. 37% Solution in Water
7647-01-0,7732-18-5
1.1800g/mL
Plastic bottle
1.18
1.9 mPa.s (15°C)
Solution, Free From Suspended Matter or Sediment
MFCD00011324 MFCD00792839
04,450; 05,533; 06,283
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrogen chloride
313
36.46
ACS Reagent

Chemical Identifiers

7647-01-0
36.46
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
Cl
ClH
MFCD00011324 MFCD00792839
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
CHEBI:17883
Videos
Safety and Handling

Safety and Handling

May cause respiratory irritation, May be corrosive to metals, Causes severe skin burns and eye damage

Specific target organ toxicity after single exposure (category 3), Corrosive to metals (category 1), Skin corrosion/irritation (category 1)

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only