missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hydrochloric acid, ACS reagent, ca. 37% solution in water, Thermo Scientific Chemicals

$75.55 - $925.25

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
SMILES Cl
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC423795000
SDS View Documents
Thermo Scientific Chemicals
423795000
500 mL Glass Bottle
Each for $75.55
 
AC423795001
SDS View Documents
Thermo Scientific Chemicals
423795001
500 mL Plastic bottle
Each for $80.61
 
AC423790025
SDS Accessories View Documents
Thermo Scientific Chemicals
423790025
2.5 L EcoSafPak™ Glass Bottle
Each for $171.31
 
AC423790251
SDS View Documents
Thermo Scientific Chemicals
423790251
25 L Plastic drum
Each for $925.25
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7647-01-0
36.46
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
Cl
ClH
MFCD00011324 MFCD00792839
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
CHEBI:17883

Specifications

7647-01-0,7732-18-5
1ppm max.
0.0005% max.
ClH
HCl
1789
15, 4818
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrogen chloride
313
36.45
ACS Reagent
1.1800g/mL
Glass Bottle
500 mL
Solution, Free From Suspended Matter or Sediment
MFCD00011324 MFCD00792839
04,450; 05,533; 06,283
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
Cl
36.46
CHEBI:17883
36.5 to 38.0%
Hydrochloric acid, ca. 37% Solution in Water
Videos
Safety and Handling

Safety and Handling

May cause respiratory irritation, May be corrosive to metals, Causes severe skin burns and eye damage

Specific target organ toxicity after single exposure (category 3), Corrosive to metals (category 1), Skin corrosion/irritation (category 1)

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only