missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Hydrochloric Acid Solution, 6N (Certified), Fisher Chemical™
Standardized at 25°C to pH 8.2 against sodium hydroxide that has been standardized against NIST™ potassium acid phthalate
$264.58 - $660.50
Chemical Identifiers
CAS | 7647-01-0 |
---|---|
Molecular Formula | ClH |
Molecular Weight (g/mol) | 36.46 |
MDL Number | MFCD00011324 MFCD00792839 |
InChI Key | VEXZGXHMUGYJMC-UHFFFAOYSA-N |
Synonym | hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof |
PubChem CID | 313 |
ChEBI | CHEBI:17883 |
IUPAC Name | hydrogen chloride |
SMILES | Cl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
SA56-500
|
Thermo Fisher Scientific Formerly Fairlawn
SA56500 |
500 mL | Glass Bottle |
Each for $264.58
Case of 6 Each for $1,587.48
|
|
||||
SA56-1
|
Thermo Fisher Scientific Formerly Fairlawn
SA561 |
1 L | Glass Bottle |
Each for $350.67
Case of 6 Each for $2,104.02
|
|
||||
SA56-4
|
Thermo Fisher Scientific Formerly Fairlawn
SA564 |
4 L | Glass Bottle |
Each for $660.50
Case of 4 Each for $2,642.00
|
|
||||
Chemical Identifiers
7647-01-0 | |
36.46 | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
313 | |
hydrogen chloride |
ClH | |
MFCD00011324 MFCD00792839 | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
CHEBI:17883 | |
Cl |
Specifications
1 | |
7647-01-0,7732-18-5 | |
5.95 to 6.05 | |
LOT 84L Potassium acid phthalate | |
500 mL | |
Liquid | |
1789 | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
hydrogen chloride | |
313 | |
Certified | |
Hydrochloric Acid Solution, 6N |
-74°C | |
81.5°C | |
Glass Bottle | |
5.7mmHg at 0°C | |
ClH | |
MFCD00011324 MFCD00792839 | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
Cl | |
36.46 | |
CHEBI:17883 | |
6N |
Safety and Handling
DOTInformation : DOT Class 8, : Corrosive