missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hydrochloric Acid Solution, 6N (Certified), Fisher Chemical™ Edge
Edge

Standardized at 25°C to pH 8.2 against sodium hydroxide that has been standardized against NIST™ potassium acid phthalate

Supplier:  Thermo Fisher Scientific Formerly Fairlawn SA561

 View more versions of this product

Catalog No. SA56-1



Edge
Explore available promotions

Description

Description

Specifications

Specifications

1
-74°C
81.5°C
Glass Bottle
5.7mmHg at 0°C
ClH
MFCD00011324 MFCD00792839
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
Cl
36.46
CHEBI:17883
6N
Hydrochloric Acid Solution, 6N
7647-01-0,7732-18-5
5.95 to 6.05
LOT 84L Potassium acid phthalate
1 L
Liquid
1789
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrogen chloride
313
Certified

Chemical Identifiers

7647-01-0
36.46
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
hydrogen chloride
ClH
MFCD00011324 MFCD00792839
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
CHEBI:17883
Cl
Videos
Safety and Handling

Safety and Handling

DOTInformation : DOT Class 8, : Corrosive

SDS Documents

Documents

Promotions

Promotions