missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hydrocinnamic acid, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 159101000

 View more versions of this product

Catalog No. AC159101000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Hydrocinnamic acid
501-52-0
1.0700g/mL
>112°C
98.5% min. (GC)
C9H10O2
MFCD00002771
09, 508
15, 4822
Solubility in water: 0.5% in water; more soluble in hot water. Other solubilities: soluble in glacial acetic acid,soluble in alcohol,benzene,ether,chloroform,,petroleum ether and carbon disulfide
C1=CC=C(C=C1)CCC(=O)O
150.18
CHEBI:28631
99%
99%
47.0°C to 50.0°C
279.0°C to 281.0°C
Authentic
Glass bottle
C6H5CH2CH2CO2H
100 g
1.07
hydrocinnamic acid, 3-phenylpropionic acid, benzylacetic acid, benzenepropanoic acid, dihydrocinnamic acid, benzenepropionic acid, phenylpropanoic acid, 3-phenyl-propionic acid, phenylpropanoate, beta-phenylpropionic acid
XMIIGOLPHOKFCH-UHFFFAOYSA-N
3-phenylpropanoic acid
107
150.18
Liquid

Chemical Identifiers

501-52-0
150.18
XMIIGOLPHOKFCH-UHFFFAOYSA-N
107
3-phenylpropanoic acid
C9H10O2
MFCD00002771
hydrocinnamic acid, 3-phenylpropionic acid, benzylacetic acid, benzenepropanoic acid, dihydrocinnamic acid, benzenepropionic acid, phenylpropanoic acid, 3-phenyl-propionic acid, phenylpropanoate, beta-phenylpropionic acid
CHEBI:28631
C1=CC=C(C=C1)CCC(=O)O
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 207-924-5

RTECSNumber : DA8600000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only