missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hydrogen bromide, pure, 33 wt% solution in glacial acetic acid, Thermo Scientific Chemicals

Hydrogen bromide, 31 to 34 wt% (HBr), BrH, CAS Number-10035-10-6, 64-19-7, acide bromhydrique, acido bromidrico, anhydrous hydrobromic acid, bromowodor, bromowodor polish, bromwasserstoff, broomwaterstof, hydrobromic acid, hydrobromide, hydrogen bromide, 2.5L, Vapor Pressure: 300mmHg at 20 deg.C

Supplier:  Thermo Scientific Chemicals 123180010

 View more versions of this product

Catalog No. AC123180010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
Specifications

Specifications

Hydrogen bromide
10035-10-6,64-19-7
Fuming Liquid
BrH
MFCD00011323
01,450; 02,214; 03,154; 04,249; 05,332; 06,282; 10,200; 15,273
09, 4665
CPELXLSAUQHCOX-UHFFFAOYSA-N
bromane
260
80.9
Pure
1.42
1.42 g/mL
Pure
Yellow
1 L
HBr
2922
39°C
hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide
Br
80.91
CHEBI:47266
31 to 34 wt% (HBr)
Glass Bottle
300mmHg at 20°C

Chemical Identifiers

10035-10-6
80.91
CPELXLSAUQHCOX-UHFFFAOYSA-N
260
Br
BrH
MFCD00011323
hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide
CHEBI:47266
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor. May cause respiratory irritation. Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only