Learn More
Hydrogen bromide, pure, 33 wt% solution in glacial acetic acid, Thermo Scientific Chemicals
Hydrogen bromide, 31 to 34 wt% (HBr), BrH, CAS Number-10035-10-6, 64-19-7, acide bromhydrique, acido bromidrico, anhydrous hydrobromic acid, bromowodor, bromowodor polish, bromwasserstoff, broomwaterstof, hydrobromic acid, hydrobromide, hydrogen bromide, 2.5L, Vapor Pressure: 300mmHg at 20 deg.C
$350.02 - $807.17
Chemical Identifiers
CAS | 10035-10-6 |
---|---|
Molecular Formula | BrH |
Molecular Weight (g/mol) | 80.91 |
MDL Number | MFCD00011323 |
InChI Key | CPELXLSAUQHCOX-UHFFFAOYSA-N |
Synonym | hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide |
PubChem CID | 260 |
ChEBI | CHEBI:47266 |
SMILES | Br |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC123180010
|
Thermo Scientific Chemicals
123180010 |
1 L | Glass Bottle |
Each for $350.02
|
|
||||
AC123180025
|
Thermo Scientific Chemicals
123180025 |
2.5 L | Glass bottle |
Each for $807.17
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentChemical Identifiers
10035-10-6 | |
80.91 | |
CPELXLSAUQHCOX-UHFFFAOYSA-N | |
260 | |
Br |
BrH | |
MFCD00011323 | |
hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide | |
CHEBI:47266 |
Specifications
10035-10-6,64-19-7 | |
Fuming Liquid | |
BrH | |
MFCD00011323 | |
01,450; 02,214; 03,154; 04,249; 05,332; 06,282; 10,200; 15,273 | |
09, 4665 | |
CPELXLSAUQHCOX-UHFFFAOYSA-N | |
bromane | |
260 | |
80.9 | |
Pure | |
1.42 | |
1.42 g/mL |
Yellow | |
1 L | |
HBr | |
2922 | |
39°C | |
hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide | |
Br | |
80.91 | |
CHEBI:47266 | |
31 to 34 wt% (HBr) | |
Glass Bottle | |
300mmHg at 20°C | |
Hydrogen bromide, Pure |
Safety and Handling
GHS H Statement
Flammable liquid and vapor. May cause respiratory irritation. Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
Danger
RUO – Research Use Only