missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Hydrogen Peroxide, 3%, BAKER ANALYZED™ Reagent, J.T. Baker™
High quality chemicals for laboratory and specialized industrial use
Supplier: Avantor 218003
Specifications
Hydrogen Peroxide | |
Poly Bottle | |
4 L | |
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone | |
OO | |
34.014 | |
34.01 | |
BAKER ANALYZED™ Reagent |
7722-84-1 | |
H2O2 | |
1L = 1.00kg | |
MHAJPDPJQMAIIY-UHFFFAOYSA-N | |
hydrogen peroxide | |
CHEBI:16240 | |
3% |
Chemical Identifiers
7722-84-1 | |
34.014 | |
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone | |
hydrogen peroxide |
H2O2 | |
MHAJPDPJQMAIIY-UHFFFAOYSA-N | |
CHEBI:16240 | |
OO |