missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hydrogen Peroxide, 3%, BAKER ANALYZED™ Reagent, J.T. Baker™

High quality chemicals for laboratory and specialized industrial use

$6440.33 - $9175.44

Chemical Identifiers

CAS 7722-84-1
Molecular Formula H2O2
Molecular Weight (g/mol) 34.014
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
Synonym oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
ChEBI CHEBI:16240
IUPAC Name hydrogen peroxide
SMILES OO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
02003182
View Documents
Avantor
218001
500 mL HDPE Pressure Bottom Poly Bottle
Case of 12 Each for $9,175.44
 
02003181
View Documents
Avantor
218003
4 L Poly Bottle
Case of 4 Each for $6,440.33
 
Description

Description

Specifications

Chemical Identifiers

7722-84-1
34.014
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
hydrogen peroxide
H2O2
MHAJPDPJQMAIIY-UHFFFAOYSA-N
CHEBI:16240
OO

Specifications

7722-84-1
H2O2
1L = 1.00kg
MHAJPDPJQMAIIY-UHFFFAOYSA-N
hydrogen peroxide
CHEBI:16240
3%
Hydrogen Peroxide
HDPE Pressure Bottom Poly Bottle
500 mL
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
OO
34.014
34.01
BAKER ANALYZED™ Reagent
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'