missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hydrogen Peroxide, 3%, BAKER ANALYZED™ Reagent, J.T. Baker™

High quality chemicals for laboratory and specialized industrial use

Supplier:  Avantor 218001

 View more versions of this product

Catalog No. 02003182

Explore available promotions

Description

Description

Specifications

Specifications

Hydrogen Peroxide
HDPE Pressure Bottom Poly Bottle
500 mL
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
OO
34.014
34.01
BAKER ANALYZED™ Reagent
7722-84-1
H2O2
1L = 1.00kg
MHAJPDPJQMAIIY-UHFFFAOYSA-N
hydrogen peroxide
CHEBI:16240
3%

Chemical Identifiers

7722-84-1
34.014
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
hydrogen peroxide
H2O2
MHAJPDPJQMAIIY-UHFFFAOYSA-N
CHEBI:16240
OO
Videos
SDS
Documents

Documents

Promotions

Promotions