missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Hydrogen peroxide 30%, Suprapur™, MilliporeSigma™
Supplier: Technidata 1072980500
Specifications
107°C (1013Â hPa) | |
7722-84-1 | |
500 mL | |
Perhydrol | |
OO | |
34.01 | |
18Â hPa (20°C) |
-26°C | |
2 to 4 (H2O, 20°C) | |
H2O2 | |
MHAJPDPJQMAIIY-UHFFFAOYSA-N | |
peroxol | |
Plastic Bottle | |
1.11Â g/cm3 (at 20°C) |
Chemical Identifiers
7722-84-1 | |
H2O2 | |
MHAJPDPJQMAIIY-UHFFFAOYSA-N | |
OO |
Safety and Handling
Recommended Storage : Store at 5 to 30°C