missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hydrogen peroxide 30%, Suprapur™, MilliporeSigma™

$477.45 - $1447.08

Chemical Identifiers

CAS 7722-84-1
Molecular Formula H2O2
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
SMILES OO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
M1072981000
View Documents
Technidata
1072981000
1 L Plastic Bottle N/A
N/A
Due to product restrictions, we cannot sell this product online.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
M1072980500
View Documents
Technidata
1072980500
500 mL Plastic Bottle N/A
N/A
Due to product restrictions, we cannot sell this product online.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

7722-84-1
H2O2
MHAJPDPJQMAIIY-UHFFFAOYSA-N
OO

Specifications

Colorless
30%
H2;O2
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
MHAJPDPJQMAIIY-UHFFFAOYSA-N
hydrogen peroxide
784
Suprapur
18 hPa (20°C)
Liquid
7722-84-1
H2O2
1.11g/cm3 (20°C)
Soluble (20°C)
OO
34.014
CHEBI:16240
Plastic Bottle
1 L
Hydrogen peroxide, Suprapur™
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at 5 to 30°C

SDS
missing translation for 'documents'

missing translation for 'documents'