missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hydrogen Peroxide, 35% Solution, Technical, 30-36%, Spectrum™ Chemical

$316.85 - $9842.32

Chemical Identifiers

CAS 7722-84-1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18610955
View Documents
Spectrum Chemical Mfg Cor
H1075500MLPL
500 mL Poly Bottle N/A
N/A
Due to product restrictions, we cannot sell this product online.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
18610962
View Documents
Spectrum Chemical Mfg Cor
H10754LT
4 L Poly Bottle N/A
N/A
Due to product restrictions, we cannot sell this product online.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
18610956
View Documents
Spectrum Chemical Mfg Cor
H1075200LTBL
200 L Drum N/A
N/A
Due to product restrictions, we cannot sell this product online.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

7722-84-1

Specifications

7722-84-1,7732-18-5
30 to 36%
Poly Bottle
1
Technical
500 mL
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.