Learn More
[Hydroxy(tosyloxy)iodo]benzene, 97+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 389610050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
oxidation reagentSpecifications
[Hydroxy(tosyloxy)iodo]benzene | |
97.0 | |
131.0°C to 137.0°C | |
Authentic | |
Glass bottle | |
MFCD00011547 | |
15, 5369 | |
LRIUKPUCKCECPT-UHFFFAOYSA-N | |
[hydroxy(phenyl)-$l^{3}-iodanyl] 4-methylbenzenesulfonate | |
325434 | |
97+% |
27126-76-7 | |
100.0 | |
Yellow | |
97+% | |
C13H13IO4S | |
5 g | |
hydroxy tosyloxy iodo benzene, htib, koser's reagent, phenyliodosohydroxy tosylate, koser reagent, unii-2eoa97b14a, hydroxy p-tolylsulfonyl oxy iodo benzene, hydroxy 4-methylbenzenesulfonato-o phenyliodine, iodosobenzene-i-mono-p-toluenesulfonate, hydroxy phenyl 4-methylbenzenesulfonoperoxoyl iodide | |
CC1=CC=C(C=C1)S(=O)(=O)OI(C2=CC=CC=C2)O | |
392.21 | |
392.21 | |
Crystalline Powder |
Chemical Identifiers
27126-76-7 | |
392.21 | |
LRIUKPUCKCECPT-UHFFFAOYSA-N | |
325434 | |
CC1=CC=C(C=C1)S(=O)(=O)OI(C2=CC=CC=C2)O |
C13H13IO4S | |
MFCD00011547 | |
hydroxy tosyloxy iodo benzene, htib, koser's reagent, phenyliodosohydroxy tosylate, koser reagent, unii-2eoa97b14a, hydroxy p-tolylsulfonyl oxy iodo benzene, hydroxy 4-methylbenzenesulfonato-o phenyliodine, iodosobenzene-i-mono-p-toluenesulfonate, hydroxy phenyl 4-methylbenzenesulfonoperoxoyl iodide | |
[hydroxy(phenyl)-$l^{3}-iodanyl] 4-methylbenzenesulfonate |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Warning
RUO – Research Use Only