missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hydroxychloroquine sulfate, 98%, Thermo Scientific Chemicals

Hydroxychloroquine sulfate, CAS # 747-36-4, is a chemical compound with antiparasitic, antiviral, and anti-rheumatic activities.

Supplier:  Thermo Scientific Chemicals 263011000

 View more versions of this product

Catalog No. AC263011000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Hydroxychloroquine sulfate shows antimalarial activity against malaria parasites mainly in the intra-erythrocytic phase of its life cycle. It acts by inhibiting the heme polymerase inside the food vacuole of the parasites
  • It shows an inhibitory activity in autophagy preventing the lysosome fusion with the autophagosome
  • Some studies demonstrated that hydroxychloroquine sulfate, due to its antiviral activity can restrict HIV infection in vitro

Applications

  • Hydroxychloroquine sulfate displays antiviral and anti-rheumatic activities
  • In vitro and in vivo experiments have been performed to investigate the role of hydroxychloroquine sulfate in the treatment of SARS-CoV-2 disease
  • Hydroxychloroquine sulfate can be used in immunofluorescence assays to evaluate the antiviral mechanisms in dengue virus-2 infection. In this disease, this compound can activate the innate immune response
Specifications

Specifications

Hydroxychloroquine sulfate
240.0°C
2% max. (105°C, 2 hrs.)
0.98
C18H26ClN3O·H2SO4
100 g
hydroxychloroquine sulfate, hydroxychloroquine sulphate, ercoquin, plaquinol, toremonil, 2-4-7-chloroquinolin-4-yl amino pentyl ethyl amino ethanol sulfate, hydroxychloroquine sulfate usp, plaquenil tn, hydroxychloroquine; sulfuric acid, dsstox_cid_27788
JCBIVZZPXRZKTI-UHFFFAOYSA-N
2-[4-[(7-chloroquinolin-4-yl)amino]pentyl-ethylamino]ethanol;sulfuric acid
12947
98%
747-36-4
White
Authentic
Glass bottle
MFCD00078203
15, 4857
Solubility in water: soluble. Other solubilities: practically insoluble in alcohol,chloroform,,ether,acetone and acetic acid
CCN(CCCC(C)NC1=C2C=CC(=CC2=NC=C1)Cl)CCO.OS(=O)(=O)O
433.96
433.96
Crystalline Powder

Chemical Identifiers

747-36-4
433.96
JCBIVZZPXRZKTI-UHFFFAOYSA-N
12947
CCN(CCCC(C)NC1=C2C=CC(=CC2=NC=C1)Cl)CCO.OS(=O)(=O)O
C18H26ClN3O·H2SO4
MFCD00078203
hydroxychloroquine sulfate, hydroxychloroquine sulphate, ercoquin, plaquinol, toremonil, 2-4-7-chloroquinolin-4-yl amino pentyl ethyl amino ethanol sulfate, hydroxychloroquine sulfate usp, plaquenil tn, hydroxychloroquine; sulfuric acid, dsstox_cid_27788
2-[4-[(7-chloroquinolin-4-yl)amino]pentyl-ethylamino]ethanol;sulfuric acid
Videos
Safety and Handling

Safety and Handling

Causes serious eye irritation, Harmful if swallowed

Serious eye damage/eye irritation (category 2), Acute toxicity (category 4)

EINECSNumber : 212-019-3

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only