missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Hydroxychloroquine sulfate, 98%, Thermo Scientific Chemicals
Hydroxychloroquine sulfate, CAS # 747-36-4, is a chemical compound with antiparasitic, antiviral, and anti-rheumatic activities.
$403.53 - $1244.90
Chemical Identifiers
CAS | 747-36-4 |
---|---|
Molecular Formula | C18H26ClN3O·H2SO4 |
Molecular Weight (g/mol) | 433.96 |
MDL Number | MFCD00078203 |
InChI Key | JCBIVZZPXRZKTI-UHFFFAOYSA-N |
Synonym | hydroxychloroquine sulfate, hydroxychloroquine sulphate, ercoquin, plaquinol, toremonil, 2-4-7-chloroquinolin-4-yl amino pentyl ethyl amino ethanol sulfate, hydroxychloroquine sulfate usp, plaquenil tn, hydroxychloroquine; sulfuric acid, dsstox_cid_27788 |
PubChem CID | 12947 |
IUPAC Name | 2-[4-[(7-chloroquinolin-4-yl)amino]pentyl-ethylamino]ethanol;sulfuric acid |
SMILES | CCN(CCCC(C)NC1=C2C=CC(=CC2=NC=C1)Cl)CCO.OS(=O)(=O)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC263010250
|
Thermo Scientific Chemicals
263010250 |
25 g | Glass bottle |
Each for $403.53
|
|
||||
AC263011000
|
Thermo Scientific Chemicals
263011000 |
100 g | Glass bottle |
Each for $1,244.90
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
- Hydroxychloroquine sulfate shows antimalarial activity against malaria parasites mainly in the intra-erythrocytic phase of its life cycle. It acts by inhibiting the heme polymerase inside the food vacuole of the parasites
- It shows an inhibitory activity in autophagy preventing the lysosome fusion with the autophagosome
- Some studies demonstrated that hydroxychloroquine sulfate, due to its antiviral activity can restrict HIV infection in vitro
Applications
- Hydroxychloroquine sulfate displays antiviral and anti-rheumatic activities
- In vitro and in vivo experiments have been performed to investigate the role of hydroxychloroquine sulfate in the treatment of SARS-CoV-2 disease
- Hydroxychloroquine sulfate can be used in immunofluorescence assays to evaluate the antiviral mechanisms in dengue virus-2 infection. In this disease, this compound can activate the innate immune response
Chemical Identifiers
747-36-4 | |
433.96 | |
JCBIVZZPXRZKTI-UHFFFAOYSA-N | |
12947 | |
CCN(CCCC(C)NC1=C2C=CC(=CC2=NC=C1)Cl)CCO.OS(=O)(=O)O |
C18H26ClN3O·H2SO4 | |
MFCD00078203 | |
hydroxychloroquine sulfate, hydroxychloroquine sulphate, ercoquin, plaquinol, toremonil, 2-4-7-chloroquinolin-4-yl amino pentyl ethyl amino ethanol sulfate, hydroxychloroquine sulfate usp, plaquenil tn, hydroxychloroquine; sulfuric acid, dsstox_cid_27788 | |
2-[4-[(7-chloroquinolin-4-yl)amino]pentyl-ethylamino]ethanol;sulfuric acid |
Specifications
747-36-4 | |
White | |
Authentic | |
Glass bottle | |
MFCD00078203 | |
15, 4857 | |
Solubility in water: soluble. Other solubilities: practically insoluble in alcohol,chloroform,,ether,acetone and acetic acid | |
CCN(CCCC(C)NC1=C2C=CC(=CC2=NC=C1)Cl)CCO.OS(=O)(=O)O | |
433.96 | |
433.96 | |
Crystalline Powder |
240.0°C | |
2% max. (105°C, 2 hrs.) | |
0.98 | |
C18H26ClN3O·H2SO4 | |
25 g | |
hydroxychloroquine sulfate, hydroxychloroquine sulphate, ercoquin, plaquinol, toremonil, 2-4-7-chloroquinolin-4-yl amino pentyl ethyl amino ethanol sulfate, hydroxychloroquine sulfate usp, plaquenil tn, hydroxychloroquine; sulfuric acid, dsstox_cid_27788 | |
JCBIVZZPXRZKTI-UHFFFAOYSA-N | |
2-[4-[(7-chloroquinolin-4-yl)amino]pentyl-ethylamino]ethanol;sulfuric acid | |
12947 | |
98% | |
Hydroxychloroquine sulfate |
Safety and Handling
Causes serious eye irritation, Harmful if swallowed
Serious eye damage/eye irritation (category 2), Acute toxicity (category 4)
EINECSNumber : 212-019-3
RUO – Research Use Only