missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Hydroxypropyl-β-cyclodextrin, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 297565000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagentSpecifications
Hydroxypropyl-β-cyclodextrin | |
278.0°C | |
Authentic | |
Plastic bottle | |
MFCD16621721 | |
Solubility in water: soluble. Other solubilities: methanol: >330g/l (25°C),chloroform: < 10g/l (25°C),dmso: >330g/l (25°C) | |
97% |
128446-35-5 | |
White to Yellow | |
2% max. Residual beta-cyclodextrin (On dry substance) (HPLC) | |
C44H75O36 | |
500 g | |
1180.05 | |
Powder |
Chemical Identifiers
128446-35-5 | |
C44H75O36 | |
1180.05 | |
MFCD16621721 |
RUO – Research Use Only