missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hydroxypropyl-β-cyclodextrin, 97%, Thermo Scientific Chemicals

$282.79 - $2197.81

Chemical Identifiers

CAS 128446-35-5
Molecular Formula C44H75O36
Molecular Weight (g/mol) 1180.05
MDL Number MFCD16621721
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC297560250
SDS View Documents
Thermo Scientific Chemicals
297560250
25 g Glass Bottle
Each for $282.79
 
AC297561000
SDS View Documents
Thermo Scientific Chemicals
297561000
100 g Glass Bottle
Each for $761.10
 
AC297565000
SDS View Documents
Thermo Scientific Chemicals
297565000
500 g Plastic bottle
Each for $2,197.81
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent
Specifications

Chemical Identifiers

128446-35-5
C44H75O36
1180.05
MFCD16621721

Specifications

128446-35-5
White to Yellow
2% max. Residual beta-cyclodextrin (On dry substance) (HPLC)
C44H75O36
25 g
1180.05
Powder
278.0°C
Authentic
Glass Bottle
MFCD16621721
Solubility in water: soluble. Other solubilities: methanol: >330g/l (25°C),chloroform: < 10g/l (25°C),dmso: >330g/l (25°C)
97%
Hydroxypropyl-β-cyclodextrin
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only