missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hydroxypropyl-β-cyclodextrin, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 297560250

 View more versions of this product

Catalog No. AC297560250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent
Specifications

Specifications

Hydroxypropyl-β-cyclodextrin
278.0°C
Authentic
Glass Bottle
MFCD16621721
Solubility in water: soluble. Other solubilities: methanol: >330g/l (25°C),chloroform: < 10g/l (25°C),dmso: >330g/l (25°C)
97%
128446-35-5
White to Yellow
2% max. Residual beta-cyclodextrin (On dry substance) (HPLC)
C44H75O36
25 g
1180.05
Powder

Chemical Identifiers

128446-35-5
C44H75O36
1180.05
MFCD16621721
Videos
SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only