missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cytiva Single Cell GenomiPhi™ DNA Amplification Kit

Amplify whole genome DNA from as little as a single cell. Cytiva Lifescience™ illustra™ Single Cell GenomiPhi™ DNA Amplification Kit combines unique capabilities of Phi29 DNA polymerase with an optimized formulation to amplify genomic DNA from 1-1000 cells without background interference.

Supplier:  Cytiva 29108039

 View more versions of this product

Catalog No. 45-500-135


Explore available promotions

Description

Description

  • For whole genome amplification of genomic DNA from as little as a single cell
  • Novel clean-up process supresses background allowing sensitivity down to 1fg of input DNA
  • Simple Isothernal strand displacement (MDA) technology using Phi29 DNA polymerase
  • Amplifications can be completed in less than 3 hours with less than 15 minutes hands-on time
  • Optimized to promote complete cell lysis with uniform denaturation of DNA prior to amplification
  • Reaction time: 2 to 4 hours
  • Product average size: >10kb
Applications

For SNP arrays, Next Gen Sequencing, aCGH

Recommended Storage

Store the kit at -70°C. The enzyme mix must be stored at -70°C; all other components may be stored at -20°C


Specifications

Specifications

Store the kit at -70°C. The enzyme mix must be stored at -70°C; all other components may be stored at -20°C.
illustra™ Single Cell GenomiPhi™ DNA Amplification Kit
Single Cell GenomiPhi Reaction buffer 1 x 275μL, Single Cell GenomiPhi Enzyme mix 1 x 25μL, Single Cell GenomiPhi Amplification mix 1 x 25μL, Single Cell GenomiPhi DTT Solution 1 x 100μL, Single Cell GenomiPhi Lysis buffer 1 x 100μL and Single Cell GenomiPhi Neutralisation buffer 1 x 100μL
illustra Single Cell GenomiPhi DNA Amplification Kit
2 to 4 hours
Single Cell GenomiPhi DNA Amplification kit combines the unique capabilities of Phi29 DNA polymerase with an optimized formulation to amplify genomic DNA from 1-1000 cells without background interference.
Genomic DNA Amplification
100
>10kb
Videos
SDS
Documents

Documents

Promotions

Promotions