missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cytiva Single Cell GenomiPhi™ DNA Amplification Kit
Amplify whole genome DNA from as little as a single cell. Cytiva Lifescience™ illustra™ Single Cell GenomiPhi™ DNA Amplification Kit combines unique capabilities of Phi29 DNA polymerase with an optimized formulation to amplify genomic DNA from 1-1000 cells without background interference.
$785.00 - $3048.00
Specifications
Content And Storage | Store the kit at -70°C. The enzyme mix must be stored at -70°C; all other components may be stored at -20°C. |
---|---|
Description | Single Cell GenomiPhi DNA Amplification kit combines the unique capabilities of Phi29 DNA polymerase with an optimized formulation to amplify genomic DNA from 1-1000 cells without background interference. |
Final Product Type | illustra™ Single Cell GenomiPhi™ DNA Amplification Kit |
For Use With (Application) | Genomic DNA Amplification |
Includes | Single Cell GenomiPhi Reaction buffer 1 x 275μL, Single Cell GenomiPhi Enzyme mix 1 x 25μL, Single Cell GenomiPhi Amplification mix 1 x 25μL, Single Cell GenomiPhi DTT Solution 1 x 100μL, Single Cell GenomiPhi Lysis buffer 1 x 100μL and Single Cell GenomiPhi Neutralisation buffer 1 x 100μL |
Includes: Single Cell GenomiPhi Reaction buffer 1 x 275µL, Single Cell GenomiPhi Enzyme mix 1 x 25µL, Single Cell GenomiPhi Amplification mix 1 x 25µL, Single Cell GenomiPhi DTT Solution 1 x 100µL, Single Cell GenomiPhi Lysis buffer 1 x 100µL and Single Cell GenomiPhi Neutralisation buffer 1 x 100µL
Catalog Number | Mfr. No. | No. of Reactions | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | No. of Reactions | Price | Quantity | |||||
45-500-134
|
Cytiva
29108107 |
25 |
Each for $785.00
|
|
|||||
45-500-135
|
Cytiva
29108039 |
100 |
Each for $3,048.00
|
|
|||||
Description
- For whole genome amplification of genomic DNA from as little as a single cell
- Novel clean-up process supresses background allowing sensitivity down to 1fg of input DNA
- Simple Isothernal strand displacement (MDA) technology using Phi29 DNA polymerase
- Amplifications can be completed in less than 3 hours with less than 15 minutes hands-on time
- Optimized to promote complete cell lysis with uniform denaturation of DNA prior to amplification
- Reaction time: 2 to 4 hours
- Product average size: >10kb
For SNP arrays, Next Gen Sequencing, aCGH
Recommended StorageStore the kit at -70°C. The enzyme mix must be stored at -70°C; all other components may be stored at -20°C
Specifications
Store the kit at -70°C. The enzyme mix must be stored at -70°C; all other components may be stored at -20°C. | |
illustra™ Single Cell GenomiPhi™ DNA Amplification Kit | |
Single Cell GenomiPhi Reaction buffer 1 x 275μL, Single Cell GenomiPhi Enzyme mix 1 x 25μL, Single Cell GenomiPhi Amplification mix 1 x 25μL, Single Cell GenomiPhi DTT Solution 1 x 100μL, Single Cell GenomiPhi Lysis buffer 1 x 100μL and Single Cell GenomiPhi Neutralisation buffer 1 x 100μL | |
>10kb |
Single Cell GenomiPhi DNA Amplification kit combines the unique capabilities of Phi29 DNA polymerase with an optimized formulation to amplify genomic DNA from 1-1000 cells without background interference. | |
Genomic DNA Amplification | |
illustra Single Cell GenomiPhi DNA Amplification Kit | |
2 to 4 hours |