Learn More
Imidazole-2-carboxaldehyde, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1388314
Description
Imidazole-2-carboxaldehyde is a novel protein tyrosine phosphatase 1B (PTP1B) inhibitor with an important application to treat type-2 diabetes. It is used in the preparation of tridentate Schiff-base carboxylate-containing ligands by undergoing condensation reaction with amino acids beta-alanine and 2-aminobenzoic acid. It is also involved in the study of the imidazole-directed allylation of aldimines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsImidazole-2-carboxaldehyde is a novel protein tyrosine phosphatase 1B (PTP1B) inhibitor with an important application to treat type-2 diabetes. It is used in the preparation of tridentate Schiff-base carboxylate-containing ligands by undergoing condensation reaction with amino acids beta-alanine and 2-aminobenzoic acid. It is also involved in the study of the imidazole-directed allylation of aldimines.
Solubility
Soluble in water.
Notes
Air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications
Imidazole-2-carboxaldehyde | |
∽205°C (decomposition) | |
MFCD00003544 | |
4371302 | |
imidazole-2-carboxaldehyde, 2-formylimidazole, 1h-imidazole-2-carboxaldehyde, imidazole-2-carbaldehyde, 2-imidazolecarboxaldehyde, imidazole-2-aldehyde, 1h-imidazolecarboxaldehyde, 2-formyl imidazole, imidazol-2-carbaldehyde, 2-formyl-1h-imidazole | |
XYHKNCXZYYTLRG-UHFFFAOYSA-N | |
1H-imidazole-2-carbaldehyde | |
24955 | |
97% |
10111-08-7 | |
C4H4N2O | |
25 g | |
Air sensitive | |
Soluble in water. | |
O=CC1=NC=CN1 | |
96.09 | |
96.09 |
Chemical Identifiers
10111-08-7 | |
96.09 | |
XYHKNCXZYYTLRG-UHFFFAOYSA-N | |
24955 | |
O=CC1=NC=CN1 |
C4H4N2O | |
MFCD00003544 | |
imidazole-2-carboxaldehyde, 2-formylimidazole, 1h-imidazole-2-carboxaldehyde, imidazole-2-carbaldehyde, 2-imidazolecarboxaldehyde, imidazole-2-aldehyde, 1h-imidazolecarboxaldehyde, 2-formyl imidazole, imidazol-2-carbaldehyde, 2-formyl-1h-imidazole | |
1H-imidazole-2-carbaldehyde |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only